missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ ML 204

Catalog No. 473210
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
473210 10 mg
473250 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 473210 Supplier Tocris Bioscience™ Supplier No. 4732/10
Only null left

Selective TRPC4 blocker

Blocker of TRPC4 channels (IC50 values are 0.96 and 2.6 μM in fluorescent and electrophysiological assays, respectively). Exhibits 19-fold selectivity against TRPC6 and 9-fold selectivity against TRPC5; displays no significant activity at TRPV1, TRPV3, TRPA1 and TRPM8 channels at concentrations up to 22 μM.

Chemical Identifiers

CAS 5465-86-1
Molecular Formula C15H18N2

Specifications

Quantity 10 mg
Formula Weight 226.32
Purity >99%
Molecular Weight (g/mol) 226.32
Chemical Name or Material ML 204
Recommended Storage Store at +4°C