missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ ML 190

Catalog No. 486650
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
486650 50 mg
486610 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 486650 Supplier Tocris Bioscience™ Supplier No. 4866/50
Only null left

Selective κ opioid receptor antagonist

Selective κ opioid receptor (KOP) antagonist (IC50 = 120 nM in a β-arrestin assay); displays >267-fold selectivity over μ and δ opioid receptors.

Chemical Identifiers

CAS 1355244-02-8
Molecular Formula C27H32N6O3

Specifications

Quantity 50 mg
Formula Weight 488.58
Molecular Weight (g/mol) 488.592
Chemical Name or Material ML 190
Recommended Storage Store at +4°C