missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ ML 145
Description
Selective antagonist for the GPR35 orphan receptor GPCR (IC50 = 20.1 nM). Over 1000-fold more selective for GPR35 compared to GPR55 antagonists.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 482.57 |
| Purity | >98% |
| Molecular Weight (g/mol) | 482.569 |
| Chemical Name or Material | ML 145 |
Safety and Handling
| Recommended Storage | Store at -20°C |