missing translation for 'onlineSavingsMsg'
Learn More

Mixed Calibration Standard 3A For Environmental Analysis, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC MIXSTD3A500

 View more versions of this product

Catalog No. MIXSTD3A500

Only null left
Explore available promotions

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
P100μg/mL
Ce20μg/mL
Co20μg/mL
V20μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Mixed Calibration Standard 3A
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
5% HNO3
Environmental
Triple-leached LDPE bottle
AA, ICP
Certified Reference Material
500 mL
Components in 5% HNO3:
Co    •20μg/mL
P    •100μg/mL
V    •20μg/mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

Product intended for laboratory use only