missing translation for 'onlineSavingsMsg'
Learn More

Mixed Calibration Standard 1A For Environmental Analysis, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC MIXSTD1A100

 View more versions of this product

Catalog No. MIXSTD1A100


Only null left
Explore available promotions

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
As100μg/mL
Ca100μg/mL
Sb50μg/mL
Se50μg/mL
B20μg/mL
Cd20μg/mL
Cu20μg/mL
Mn20μg/mL
Ba10μg/mL
Ag5μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Mixed Calibration Standard 1A
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Nitric Acid/Trace Tartaric Acid/Trace Hydrogen Fluoride
Environmental
Triple-leached LDPE bottle
AA, ICP
Certified Reference Material
125 mL
Components in 5% HNO3:
Sb    •50μg/mL
As    •100μg/mL
Ba    •10μg/mL
B    •10μg/mL
Cd    •20μg/mL
Ca    •100μg/mL
Cu    •20μg/mL
Mn    •20μg/mL
Se    •50μg/mL
Ag    •5μg/mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

Product intended for laboratory use only