missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mixed Calibration Standard 1 For Environmental Analysis, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Supplier: SPEX CertiPrep Group LLC MIXSTD1100
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| Pb | 500μg/mL |
| Se | 200μg/mL |
| Cd | 150μg/mL |
| Zn | 150μg/mL |
| Mn | 100μg/mL |
| Be | 50μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
| Mixed Calibration Standard 1 | |
| Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
| AA, ICP | |
| Certified Reference Material | |
| 125 mL | |
| Components in 2% HNO3:Be •50μg/mL Cd •150μg/mL Pb •500μg/mL Mn •100μg/mL Se •200μg/mL Zn •150μg/mL |
| Triple-leached LDPE bottle | |
| US EPA Method 200.7 (Rev. 3.3) and SW-846, Method 6010 (3rd Ed.) | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| 2% HNO3 | |
| Environmental |
Product intended for laboratory use only