missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mixed Calibration Standard 1 For Environmental Analysis, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
$768.69 - $768.69
Specifications
| Chemical Name or Material | Mixed Calibration Standard 1 |
|---|---|
| Packaging | Triple-leached LDPE bottle |
| Concentration or Composition Notes | Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. |
| Technique | AA, ICP |
| For Use With (Equipment) | AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
MIXSTD1-100
|
SPEX CertiPrep Group LLC
MIXSTD1100 |
125 mL |
Each for $768.69
|
|
|||||
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| Pb | 500μg/mL |
| Se | 200μg/mL |
| Cd | 150μg/mL |
| Zn | 150μg/mL |
| Mn | 100μg/mL |
| Be | 50μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
| Mixed Calibration Standard 1 | |
| Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| 2% HNO3 | |
| Components in 2% HNO3:Be •50μg/mL Cd •150μg/mL Pb •500μg/mL Mn •100μg/mL Se •200μg/mL Zn •150μg/mL |
| Triple-leached LDPE bottle | |
| AA, ICP | |
| Certified Reference Material | |
| Environmental |
Product intended for laboratory use only