missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mirus Bio™ TransIT-X2™ Dynamic Delivery System
Non-liposomal polymeric system for efficient transfection of plasmid DNA and siRNA/miRNA. Transfections do not require medium changes and can be performed in serum-containing medium.
Supplier: Mirus Bio™ MIR6000
Description
- Efficiency: Provides exceptional broad spectrum transfection
- Versatility: Facilitates cutting edge delivery of plasmid DNA, siRNA and miRNA
- Functions on a novel, non-liposomal polymeric technology
Specifications
| -20°C | |
| Tube | |
| Chemical Transfection | |
| Transfection Reagent | |
| Sterile |
| Established Cell Lines, Stem Cells, Primary Cells, Hard-to-Transfect Cells | |
| None (Promoterless) | |
| High efficiency delivery of plasmid DNA and siRNA/miRNA | |
| 1.5 mL |