missing translation for 'onlineSavingsMsg'
Learn More

Mirus Bio™ TransIT-X2™ Dynamic Delivery System

Catalog No. p-4654944
Encompass
Change view
Click to view available options
Quantity:
0.3 mL
0.75 mL
1.5 mL
5 x 1.5 mL
10 x 1.5 mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MIR6003 0.3 mL
MIR6004 0.75 mL
MIR6000 1.5 mL
MIR6005 5 x 1.5 mL
MIR6006 10 x 1.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MIR6003 Supplier Mirus Bio™ Supplier No. MIR6003
Only null left

Non-liposomal polymeric system for efficient transfection of plasmid DNA and siRNA/miRNA. Transfections do not require medium changes and can be performed in serum-containing medium.

  • Efficiency: Provides exceptional broad spectrum transfection
  • Versatility: Facilitates cutting edge delivery of plasmid DNA, siRNA and miRNA
  • Functions on a novel, non-liposomal polymeric technology

Specifications

Content And Storage -20°C
Cell Type Established Cell Lines, Stem Cells, Primary Cells, Hard-to-Transfect Cells
Format Tube
Promoter None (Promoterless)
Transfection Technique Chemical Transfection
For Use With (Application) High efficiency delivery of plasmid DNA and siRNA/miRNA
Product Type Transfection Reagent
Quantity 0.3 mL
Sterility Sterile