missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Mirin, 95%
CAS: 299953-00-7 | C10H8N2O2S, C10H8N2O2S | 220.25 g/mol
Chemical Identifiers
| CAS | 299953-00-7 |
|---|---|
| Molecular Formula | C10H8N2O2S |
| Synonym | cis-5-(4-Hydroxybenzylidene)-2-imino-4-thiazolidinone; |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAJ67462LB0
|
Thermo Scientific Chemicals
J67462LB0 |
5 mg |
N/A
|
|
|||||
|
AAJ67462MF
|
Thermo Scientific Chemicals
J67462MF |
50 mg |
N/A
|
|
|||||
|
AAJ67462MA
|
Thermo Scientific Chemicals
J67462MA |
10 mg |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 299953-00-7 | |
| C10H8N2O2S | |
| cis-5-(4-Hydroxybenzylidene)-2-imino-4-thiazolidinone; |
Specifications
| Orange | |
| 299953-00-7 | |
| 220.25g/mol | |
| Moisture Sensitive | |
| YBHQCJILTOVLHD-UHFFFAOYSA-N | |
| 2-amino-5-[(4-hydroxyphenyl)methylidene]-4,5-dihydro-1,3-thiazol-4-one | |
| 95% |
| -30°C to -10°C | |
| C10H8N2O2S | |
| cis-5-(4-Hydroxybenzylidene)-2-imino-4-thiazolidinone; | |
| Solid | |
| NC1=NC(=O)C(S1)=CC1=CC=C(O)C=C1 | |
| 220.25 |
Safety and Handling
Recommended Storage : Store at -20°C; Store under Nitrogen
RUO – Research Use Only