CAS RN 299953-00-7
CAS RN 299953-00-7
IUPAC Name:
2-amino-5-[(4-hydroxyphenyl)methylidene]-4,5-dihydro-1,3-thiazol-4-one
Synonyms:
2-amino-5-[(4-hydroxyphenyl)methylidene]-1,3-thiazol-4-one
Molecular Weight (g/mol):
220.25
Molecular Formula:
C10H8N2O2S
InChi Key:
YBHQCJILTOVLHD-UHFFFAOYSA-N
SMILES:
NC1=NC(=O)C(S1)=CC1=CC=C(O)C=C1
1
–
1
of
1
results
1
Thermo Scientific Chemicals Mirin, 95%
CAS: 299953-00-7 Molecular Formula: C10H8N2O2S Synonym: cis-5-(4-Hydroxybenzylidene)-2-imino-4-thiazolidinone;