missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Mirin, 95%

Catalog No. AAJ67462MF
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67462MF 50 mg
AAJ67462LB0 5 mg
AAJ67462MA 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ67462MF Supplier Thermo Scientific Chemicals Supplier No. J67462MF

missing translation for 'validMainframeMsg'

CAS: 299953-00-7 | C10H8N2O2S, C10H8N2O2S | 220.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 299953-00-7
Molecular Formula C10H8N2O2S
Synonym cis-5-(4-Hydroxybenzylidene)-2-imino-4-thiazolidinone;

Specifications

Quantity 50 mg
Description Orange
Content And Storage -30°C to -10°C
Formula Weight 220.25g/mol
Sensitivity Moisture Sensitive
Form Solid
Assay 95%

RUO – Research Use Only