missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Mirin, 95%

CAS: 299953-00-7 | C10H8N2O2S, C10H8N2O2S | 220.25 g/mol

Supplier:  Thermo Scientific Chemicals J67462MA

 View more versions of this product

Catalog No. AAJ67462MA


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

10 mg
-30°C to -10°C
299953-00-7
220.25g/mol
Moisture Sensitive
YBHQCJILTOVLHD-UHFFFAOYSA-N
2-amino-5-[(4-hydroxyphenyl)methylidene]-4,5-dihydro-1,3-thiazol-4-one
Orange
95%
C10H8N2O2S
cis-5-(4-Hydroxybenzylidene)-2-imino-4-thiazolidinone;
Solid
NC1=NC(=O)C(S1)=CC1=CC=C(O)C=C1
220.25

Chemical Identifiers

299953-00-7
C10H8N2O2S
cis-5-(4-Hydroxybenzylidene)-2-imino-4-thiazolidinone;
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at -20°C; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only