missing translation for 'onlineSavingsMsg'
Learn More

Milnacipran Hydrochloride 98.0+%, TCI America™

Catalog No. p-7133042
Change view
Click to view available options
Quantity:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M213350MG 50 mg
M2133500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M213350MG Supplier TCI America Supplier No. M213350MG
Only null left

Chemical Identifiers

CAS 101152-94-7
Molecular Formula C15H23ClN2O
Molecular Weight (g/mol) 282.812
MDL Number MFCD00901293
InChI Key XNCDYJFPRPDERF-LKNSGBSQSA-N
Synonym dalcipran, ixel, rac-milnacipran hydrochloride, 1r,2s-rel-2 aminomethyl-n,n-diethyl-1-phenyl-cyclopropanecarboxamide, monohydrochloride, 2s-2-aminomethyl-n,n-diethyl-1-phenylcyclopropane-1-carboxamide hydrochloride
PubChem CID 45039934
IUPAC Name (2S)-2-(aminomethyl)-N,N-diethyl-1-phenylcyclopropane-1-carboxamide;hydrochloride
SMILES CCN(CC)C(=O)C1(CC1CN)C2=CC=CC=C2.Cl

Specifications

Melting Point 181°C
Color White-Yellow
Quantity 50 mg
UN Number 2811
Formula Weight 282.81
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Milnacipran Hydrochloride
RTECSNumber GZ1014010
TSCA No
Recommended Storage Refrigerator