Learn More
MilliporeSigma™ Millicell™ Receiver Trays
Comes with lid
Supplier: MilliporeSigma™ PSSW010R5
Description
Designed to support cell attachment, growth, and differentiation for a variety of applications
- 24-well feeder tray incorporates a tear-drop well design to reduce the incidence of air bubbles that can form under the membrane during plate assembly, ensuring that cell monolayers are getting the proper nutrients
- Single-well feeder tray utilizes baffles to reduce media leakage and contamination
Specifications
| With Lid | |
| Polystyrene | |
| Sterile | |
| 1 | |
| 5 μm |
| Clear | |
| Rectangular | |
| Receiver Tray | |
| Lid, Single Well Receiver Tray |