Learn More
MilliporeSigma™ Millicell™ Receiver Trays
Comes with lid
$161.00 - $161.00
Specifications
| Closure Included | With Lid |
|---|---|
| Color | Clear |
| Material | Polystyrene |
| Shape | Rectangular |
| Sterility | Sterile |
Description
Designed to support cell attachment, growth, and differentiation for a variety of applications
- 24-well feeder tray incorporates a tear-drop well design to reduce the incidence of air bubbles that can form under the membrane during plate assembly, ensuring that cell monolayers are getting the proper nutrients
- Single-well feeder tray utilizes baffles to reduce media leakage and contamination
Specifications
| With Lid | |
| Polystyrene | |
| Sterile |
| Clear | |
| Rectangular | |
| Receiver Tray |