missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Mildronate dihydrate

Catalog No. AC466260010
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC466260010 1 g
AC466262500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC466260010 Supplier Thermo Scientific Chemicals Supplier No. 466260010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 86426-17-7 | C6H18N2O4, C6 H14 N2 O2 . 2 H2 O | 182.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 86426-17-7
Molecular Formula C6 H14 N2 O2 . 2 H2 O

Specifications

Quantity 1 g
Solubility soluble aq. soln.
Formula Weight 182.22
Physical Form Crystalline Powder
Product Type Reagent
Purity 97.5 to 102.5% (Titration with HClO4)
Chemical Name or Material Mildronate dihydrate

RUO – Research Use Only