missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MicroSEQ™ ID Sequencing Clean-up Cartridges

Catalog No. 4408228
Change view
Click to view available options
Quantity:
36 cartridges
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4408228 36 cartridges
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4408228 Supplier Applied Biosystems™ Supplier No. 4408228
Only null left

The MicroSEQ ID Sequencing Clean-up Cartridges are used for post-sequencing clean-up in the MicroSEQ ID workflow.

The MicroSEQ™ ID Sequencing Clean-up Cartridges are used for post-sequencing clean-up in the MicroSEQ™ ID workflow. The gel filtration matrix used in the cartridges provides a convenient and simple way to remove dye terminators, dNTPs, and salts, as well as residual primers from cycle sequencing reactions.

The MicroSEQ™ ID Sequencing Clean-up Cartridges can be used downstream of the following sequencing kits:

  • MicroSEQ™ 500 16S rDNA Sequencing Kit
  • MicroSEQ™ Full Gene 16S rDNA Sequencing Kit
  • MicroSEQ™ D2 rDNA Fungal Sequencing Kit

The kit consists of 36 cartridges and is ideal for low-throughput microbial identification workflows. The end products can be directly used for sequencing on any of Applied Biosystems™ genetic analyzers.

Specifications

Content And Storage Store in refrigerator (2°C to 8°C).
Format Cartridge, Spin Column
Isolation Technology Spin Column
Final Product Type DNA
For Use With (Equipment) MicroSEQ™ ID
For Use With (Application) Sequencing
High-throughput Compatibility Not High-throughput Compatible (Manual)
Product Line MicroSEQ
Quantity 36 cartridges
Purity Sequencing Grade
Shipping Condition Wet Ice
Target Sequencing Reactions
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.