missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MicroSEQ™ ID Purification Combo Kit v2.0, with cartridges

Catalog No. A37741
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A37741 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A37741 Supplier Applied Biosystems™ Supplier No. A37741

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The MicroSEQ ID Purification Combo Kit v2.0 includes reagents for post-PCR and post-sequencing clean-up in the MicroSEQ ID microbial identification workflow.

The MicroSEQ ID Purification Combo Kit v2.0 includes reagents for post-PCR and post-sequencing clean-up in the MicroSEQ ID microbial identification workflow. This version contains one tube of ExoSAP-IT Express Reagent for post-PCR clean-up and MicroSEQ ID Sequencing Clean-up Cartridges for post-sequencing clean-up.

ExoSAP-IT Express Reagent removes unincorporated dNTPs and primers from the post-amplification PCR mix in 5 minutes compared to 30 minutes with the original ExoSAP-IT Reagent while producing equivalent results. The MicroSEQ ID Sequencing Clean-up Cartridges provide a convenient and simple way to remove dye terminators, dNTPs, salts, and residual primers from cycle sequencing reactions. The end products can be used directly for sequencing on any Applied Biosystems genetic analyzer.

Comparability studies demonstrated equivalent results between the original ExoSAP-IT Reagent and the improved ExoSAP-IT Express Reagent coupled with the MicroSEQ ID Sequencing Clean-Up kit in the MicroSEQ ID workflow (see figure 1). Both ExoSAP-IT and ExoSAP-IT Express enzyme-treated PCR samples produced very similar sequencing results and the correct identification result.

ExoSAP-IT Express Reagent can be used downstream of the following PCR amplification kits:

  • MicroSEQ 500 16S rDNA PCR Kit
  • MicroSEQ Full Gene 16S rDNA PCR Kit
  • MicroSEQD2 rDNA Fungal PCR Kit

MicroSEQ ID Sequencing Clean-up Cartridges can be used downstream of the following sequencing kits:

  • MicroSEQ 500 16S rDNA Sequencing Kit
  • MicroSEQ Full Gene 16S rDNA Sequencing Kit
  • MicroSEQ D2 rDNA Fungal Sequencing Kit

Specifications

Content And Storage • ExoSAP-IT Express (100 reactions), store at -5°C to -30°C
• MicroSEQ ID Sequencing Clean-up Cartridges (36 cartridges), store at 2°C to 8°C
Format Cartridge
For Use With (Application) Sequencing
High-throughput Compatibility Manual Protocols
No. of Reactions 100 Reactions
Product Line MicroSEQ
Quantity 1 kit
Shipping Condition Approved for shipment on Wet or Dry Ice

For Research Use Only. Not for use in diagnostic procedures.