Learn More
Applied Biosystems™ MicroSEQ™ ID Ultra Sequencing Clean-up 8-Strips Kit
Description
The MicroSEQ™ ID Ultra Sequencing Clean-up 8-Strips Kit is used for post-sequencing clean-up in the MicroSEQ™ ID workflow. The gel-filtration matrix used in the strip format provides a convenient and simple way to remove dye terminators, dNTPs, salts, and residual primers from cycle sequencing reactions.
The MicroSEQ ID Ultra Sequencing Clean-up 8-Strips Kit can be used downstream of the following sequencing kits:
- MicroSEQ™ 500 16S rDNA Sequencing Kit
- MicroSEQ™ Full Gene 16S rDNA Sequencing Kit
- MicroSEQ™ D2 rDNA Fungal Sequencing Kit
The kit consists of twelve 8-well strips and a 96-well receiver plate to accommodate 96 sequencing samples. It is ideal for low- to medium-throughput microbial-identification workflows. The end products can be used directly for sequencing on any Applied Biosystems™ genetic analyzers.
Specifications
Specifications
| Content And Storage | • 12 x 8-strip clean-up tubes • 2 x 96-well waste plate • 2 x 96-well receiver plate • 1 x 8-strip balancing strip |
| Format | Strip |
| Sample Type | Sequencing Products |
| For Use With (Application) | Sequencing |
| High-throughput Compatibility | Manual Protocols |
| No. of Samples | 96 Samples |
| Product Line | MicroSEQ™ ID |
| Quantity | 96 samples |
| Target | Fluorescent Dye Removal |
Frequently Asked Questions (FAQs)
The product is guaranteed stable for one year when stored properly at 4 degrees C.
The MicroSEQ ID Ultra Sequencing Clean-up 8-Strips Kit is sufficient for 96 sequencing samples.
Here the kits with the catalog numbers:
- MicroSEQ ID Ultra Sequencing Clean-up 8-Strips Kit (Cat. No. A33246)
- MicroSEQ ID Ultra Sequencing Clean-up Plates Kit (Cat. No.A33245)
- MicroSEQ ID Ultra Sequencing Clean-up 8-Strips Refill Kit (Cat. No. A33256)
The kit consists of twelve 8-well strips and a 96-well receiver plate to accommodate 96 sequencing samples.
Strips should be used within 2 weeks of opening. We recommend returning unused strips from the 96-well plate holder to the foil package, sealing, and storing at 4 degrees C.