missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ MicroSEQ™ Salmonella spp. Detection Kit
Specifications
Specifications
| Content And Storage | This kit includes 12 8-tube strips containing assay beads (store at 2 to 8°C in the dark), optical 8-tube strips and 8-cap strips (store at 22 to 28°C), and a Negative Control (store at 2 to 8°C). These reagents are sufficient for 96 reactions. |
| Detection Method | Primer-probe |
| PCR Method | qPCR |
| Polymerase | AmpliTaq Fast DNA Polymerase |
| Reaction Speed | Fast |
| For Use With (Equipment) | 7500 Fast System, StepOne™, Fast Mode, StepOnePlus™, StepOnePlus™, Fast Mode |
| Kit Type | MicroSEQ Salmonella spp. Detection Kit |
| Passive Reference Dye | ROX (Pre-mixed) |
| Product Line | MicroSEQ |
| Quantity | 1 kit |
| Show More |
Frequently Asked Questions (FAQs)
Which sample preparation kit is compatible with the MicroSEQ Salmonella spp. Detection Kit?
The MicroSEQ Salmonella spp. Detection Kit is optimized for use with PrepSEQ Sample Preparation Kits.