missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MicroSEQ™ D2 rDNA Fungal PCR Kit
GREENER_CHOICE

Catalog No. 4349153
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4349153 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4349153 Supplier Applied Biosystems™ Supplier No. 4349153
Get an Extra Analytical Consumable with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This kit is the PCR component of the MicroSEQ™ Fungal Identification System, which provides an easy-to-use DNA sequence-based method to identify most fungi.

This kit is the PCR component of the MicroSEQ™ Fungal Identification System, which provides an easy-to-use DNA sequence-based method to identify most fungi. A protocol and Quick Reference Card are not included.

  • Sequence-based measurement ensures you get reproducible results in less than 24 hours.
  • Growth requirement-independent measurement enables you to identify fungi using objective criteria.
  • Universal domain-level primers eliminate up-front optimization.
  • MicroSEQ™ ID Analysis Software ensures accurate species identification.
  • Customizable libraries give you the flexibility to create new libraries and search validated libraries that contain the most frequently encountered organisms.
  • Requires minimal training; same work flow as the MicroSEQ™ Full Gene 16S rDNA Bacterial Identification Kit.

Specific and Easy to Use

Based on phylogenetic analysis of the D2 region of large-subunit ribosomal RNA gene sequences, this easy-to-use kit enables you to identify even the most difficult fungi, yeast, and oomycete plant pathogens. To ensure accurate and specific results, the kit's software compares your samples to a database of D2 sequences from over 1,000 validated type species.

The MicroSEQ™ ID System: A Complete, Integrated Solution

Streamlining every step in microbial identification, Applied Biosystems MicroSEQ™ System combines the advantages of MicroSEQ™ ID Software with easy PrepMan Ultra sample preparation reagents and protocols, MicroSEQ™ bacterial and fungal application kits, and industry-leading thermal cyclers and sequencing systems. To ensure optimal performance and system-wide integration, we develop and test all components of the system together.

Specifications

Content And Storage The kit contains reagents sufficient for 50 PCR reactions:
PCR Master Mix
Positive Control DNA
Negative Control (water)
After first use, store the kit in a PCR clean room at 2–8 °C
Detection Method Primer-probe
Technique PCR Amplification, SBT (Sequence-Based Typing)
For Use With (Equipment) Veriti Thermal Cycler, 9800 Fast Thermal Cycler
Libraries Fungal Library (> 1100 species), Customizable
Product Line MicroSEQ
Product Type PCR Kit
Quantity 1 Kit
Shipping Condition Dry Ice
Software MicroSEQ™ ID Analysis Software
Target D2 region of rRNA gene
Target Organism Class Fungi
For Use With (Application) Microbial Detection
No. of Reactions 50 Reactions (PCR)
Sample Type DNA (Genomic)
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.