Learn More
Gilson™ MICROMAN™ E Positive Displacement Pipette PROMO
The ergonomically designed MICROMAN™ E Positive Displacement Pipette, along with disposable capillary piston tips, provide the highest precision when pipetting viscous, non-aqueous liquids while protecting the pipette from contamination.
$750.78 - $750.78
Specifications
| Compatible Tips | CP Tips |
|---|---|
| Autoclavable | Autoclavable |
| Material | Polypropylene |
| Number of Channels | 1 |
| Type | Positive Displacement Pipette |
| Catalog Number | Mfr. No. | Volume (Metric) | Model | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Volume (Metric) | Model | Price | Quantity | ||||
|
FD10001GPM
|
Gilson™
FD10001GPM |
1 to 10 μL | M10E |
Each for $750.78
|
|
||||
|
FD10002GPM
|
Gilson™
FD10002GPM |
3 to 25 μL | M25E |
Each for $750.78
|
|
||||
|
FD10003GPM
|
Gilson™
FD10003GPM |
20 to 50 μL | M50E |
Each for $750.78
|
|
||||
|
FD10004GPM
|
Gilson™
FD10004GPM |
10 to 100 μL | M100E |
Each for $750.78
|
|
||||
|
FD10005GPM
|
Gilson™
FD10005GPM |
50 to 250 μL | M250E |
Each for $750.78
|
|
||||
Description
MICROMAN™ E positive displacement pipettes with capillary pistons offer highest precision when pipetting problem liquids such as oils, cosmetics, blood, glycerol, radioactive samples, RNA, DNA, and more.
- Ergonomic design to increase pipetting performance.
- Patented QuickSnap feature for Capillary Piston fitting System.
- Volume securing button to reduce risk of accidental volume change during pipetting cycles.
- Comfortable and easy to use with new handle shape and added finger hook.
- Fully visible volumeter.
- Large pushbutton for allows easy aspirate and dispense.
- For complete and rapid pipetting of viscous, volatile, dense, hot or cold liquids.
- Avoids leaks when pipetting volatile (high vapor pressure) liquids.
- Avoid cross-contamination or residual sample carry-over.
- For tips, see F148412G or F148413G.
Specifications
| CP Tips | |
| Polypropylene | |
| Positive Displacement Pipette | |
| 1 Ea. |
| Autoclavable | |
| 1 | |
| For pipetting viscous, volatile, and problem liquids | |
| Certified Quality |