missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences MicroLiter 9mm Screwthread Autosampler Vial Caps with Open Tops

MicroLiter's 9mm autosampler screw-thread vial caps with open tops are available in multiple colors.

Supplier:  DWK Life Sciences 090000K

 View more versions of this product

Catalog No. 15111009


Only null left

Description

Description

For use with MicroLiter 9mm Screwthread Autosample Vial Septa sold separately.
Specifications

Chemical Identifiers

Polypropylene
100
9 mm Autosampler Vials - MicroLiter
None
9 mm
Screw Cap

Specifications

Black
None
9 mm
Screw Cap
Polypropylene
100
9 mm Autosampler Vials - MicroLiter
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions