missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences MicroLiter 20mm Magnetic Large Opening Crimp Top Headspace Vial Seals

MicroLiter 20mm crimp top seals are available in multiple colors and septa types.

Supplier:  DWK Life Sciences 200051MLB

 View more versions of this product

Catalog No. 15111732


Only null left

Description

Description

Magnetic large opening crimp seal
Specifications

Chemical Identifiers

Steel
20 mm
Crimp Cap
No
20 mm Headspace Vials - MicroLiter

Specifications

Tan PTFE/White silicone
Steel
PTFE/Silicone
20 mm
Crimp Cap
Silver
No
1000
20 mm Headspace Vials - MicroLiter
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions