missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences MicroLiter 20mm Extended Apron Magnetic Crimp Top Headspace Vial Seals

MicroLiter 20mm crimp top seals are available in multiple colors and septa types.

$41.90

Specifications

Material Steel
Septum None
Quantity 100
Closure Size 20 mm
For Use With (Equipment) 20 mm Headspace Septa - MicroLiter
View More Specs

Products 1
Catalog Number Mfr. No. Color Price Quantity & Availability  
Catalog Number Mfr. No. Color Price Quantity & Availability  
15111639
View Documents Promotion Details
DWK Life Sciences
200000MS
Gold
Pack of 100 Each for $41.90
Only null left
 
Description

Description

Magnetic crimp seal
Specifications

Specifications

Steel
100
20 mm Headspace Septa - MicroLiter
None
20 mm
Crimp Cap
Videos
SDS
Documents

Documents