missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences MicroLiter 16mm Septa for Screwthread Headspace Vial Caps
MicroLiter septa for 16mm Screwthread Headspace Vials are constructed of PTFE, PTFE/butyl, or PTFE/silicone to suit a variety of chromatography applications.
Supplier: DWK Life Sciences 160050
Chemical Identifiers
| No | |
| 100 | |
| 16 mm | |
| 16 mm Screw Thread Headspace Vials - MicroLiter |
Specifications
| Blue | |
| PTFE/Silicone | |
| 16 mm |
| No | |
| 100 | |
| 16 mm Screw Thread Headspace Vials - MicroLiter |