missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences MicroLiter 16 mm Screw Thread Headspace Vial Caps

MicroLiter's 16 mm screw thread headspace vial caps are available in multiple septa types.

$131.90 - $3,978.78

Specifications

Color Silver
Material Steel
Product Line MicroLiter
Closure Size 16 mm
For Use With (Equipment) 16 mm Screw Thread Headspace Vials - MicroLiter
View More Specs

Products 6
Catalog Number Mfr. No. Septum Price Quantity & Availability  
Catalog Number Mfr. No. Septum Price Quantity & Availability  
15111604
View Documents Promotion Details
DWK Life Sciences
160000M
None
Pack of 100 Each for $131.90
Only null left
 
15111607
View Documents Promotion Details
DWK Life Sciences
160030M
PTFE, Butyl
Pack of 100 Each for $184.22
Only null left
 
15111609
View Documents Promotion Details
DWK Life Sciences
160050M
White PTFE, Blue Silicone
Pack of 100 Each for $412.22
Only null left
 
15111610
View Documents Promotion Details
DWK Life Sciences
160050MB
PTFE, Silicone
Pack of 1000 Each for $3,978.78
Only null left
 
15111612
View Documents Promotion Details
DWK Life Sciences
160055M
Blue PTFE, White Silicone
Pack of 100 Each for $484.00
Only null left
 
15111614
View Documents Promotion Details
DWK Life Sciences
W 160080M
FKM
Pack of 100 Each for $155.83
Only null left
 
Description

Description

Specifications

Specifications

Silver
MicroLiter
16 mm Screw Thread Headspace Vials - MicroLiter
Steel
16 mm
Screw Cap
Videos
SDS
Documents

Documents