missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences MicroLiter 16 mm Screw Thread Headspace Vial Caps

Catalog No. 15111612
Change view
Click to view available options
Septum:
Blue PTFE, White Silicone
FKM
None
PTFE, Butyl
PTFE, Silicone
White PTFE, Blue Silicone
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Septum
15111612 Blue PTFE, White Silicone
15111604 None
15111607 PTFE, Butyl
15111609 White PTFE, Blue Silicone
15111610 PTFE, Silicone
15111614 FKM
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 15111612 Supplier DWK Life Sciences Supplier No. 160055M

MicroLiter's 16 mm screw thread headspace vial caps are available in multiple septa types.

Specifications

Color Silver
Material Steel
Pre-slit No
Septum Blue PTFE, White Silicone
No. per Pack 100
Product Line MicroLiter
Closure Size 16 mm
For Use With (Equipment) 16 mm Screw Thread Headspace Vials - MicroLiter
Product Type Screw Cap