missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences MicroLiter 11 mm Amber Glass Crimp Top Autosampler Vials With Insert

MicroLiter 11 mm Crimp Top Autosampler Vials With Inserts are cleaned and packaged in a certified Class 10,000 clean room according to a designed process, developed to eliminate chromatographic errors associated with vials. Vials and inserts meet strict tolerances.

Supplier:  DWK Life Sciences W 112200101

 View more versions of this product

Catalog No. 15111590


Only null left

Description

Description

For use with Microliter 11 mm Crimp Top Autosampler Vial seals
Specifications

Chemical Identifiers

Amber
Borosilicate Glass (Type 1)
MicroLiter
Yes
12 mm
100
11 mm
32 mm

Specifications

Amber
Borosilicate Glass (Type 1)
Conical with Pulled Point
MicroLiter
Yes
12 mm
350 μL
100
11 mm
32 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions