missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Microcrystalline Cellulose, NF, BP, JP, Spectrum™ Chemical
C4578, 9004-34-6
$274.20 - $7128.27
Chemical Identifiers
| CAS | 9004-34-6 |
|---|---|
| Molecular Formula | C14H26O11 |
| Molecular Weight (g/mol) | 370.35 |
| InChI Key | PTHCMJGKKRQCBF-UHFFFAOYNA-N |
| IUPAC Name | 2-{[4,5-dihydroxy-2-(hydroxymethyl)-6-methoxyoxan-3-yl]oxy}-6-(hydroxymethyl)-5-methoxyoxane-3,4-diol |
| SMILES | COC1OC(CO)C(OC2OC(CO)C(OC)C(O)C2O)C(O)C1O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18604596
|
Spectrum Chemical Mfg Cor
C4578100GM |
100 g | Poly Bottle |
Each for $274.20
|
|
||||
|
18604597
|
Spectrum Chemical Mfg Cor
C4578500GM |
500 g | Poly Bottle |
Each for $719.90
|
|
||||
|
18604598
|
Spectrum Chemical Mfg Cor
C457825KG |
2.5 kg | Poly Pail |
Each for $2,408.53
|
|
||||
|
18604599
|
Spectrum Chemical Mfg Cor
C457812KG |
12 kg | Fiber Drum |
Each for $3,513.83
|
|
||||
|
18604600
|
Spectrum Chemical Mfg Cor
C457850KGBL |
50 kg | Fiber Drum |
Each for $7,128.27
|
|
||||
Description
Spectrum™ Chemical Microcrystalline Cellulose, NF, BP, JPChemical Identifiers
| 9004-34-6 | |
| 370.35 | |
| 2-{[4,5-dihydroxy-2-(hydroxymethyl)-6-methoxyoxan-3-yl]oxy}-6-(hydroxymethyl)-5-methoxyoxane-3,4-diol |
| C14H26O11 | |
| PTHCMJGKKRQCBF-UHFFFAOYNA-N | |
| COC1OC(CO)C(OC2OC(CO)C(OC)C(O)C2O)C(O)C1O |
Specifications
| 9004-34-6 | |
| C14H26O11 | |
| PTHCMJGKKRQCBF-UHFFFAOYNA-N | |
| 2-{[4,5-dihydroxy-2-(hydroxymethyl)-6-methoxyoxan-3-yl]oxy}-6-(hydroxymethyl)-5-methoxyoxane-3,4-diol | |
| Multi-Compendial |
| 1 | |
| 100 g | |
| COC1OC(CO)C(OC2OC(CO)C(OC)C(O)C2O)C(O)C1O | |
| 370.35 | |
| Poly Bottle |