missing translation for 'onlineSavingsMsg'
Learn More

Microcrystalline Cellulose, NF, BP, JP, Spectrum™ Chemical

Catalog No. 18604596
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604596 100 g Poly Bottle
18604597 500 g Poly Bottle
18604598 2.5 kg Poly Pail
18604599 12 kg Fiber Drum
18604600 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18604596 Supplier Spectrum Chemical Mfg Cor Supplier No. C4578100GM
Get Up to $500 in Chemicals with Purchase
View all promotions

C4578, 9004-34-6

Spectrum™ Chemical Microcrystalline Cellulose, NF, BP, JP

Chemical Identifiers

CAS 9004-34-6
Molecular Formula C14H26O11
Molecular Weight (g/mol) 370.35
InChI Key PTHCMJGKKRQCBF-UHFFFAOYNA-N
IUPAC Name 2-{[4,5-dihydroxy-2-(hydroxymethyl)-6-methoxyoxan-3-yl]oxy}-6-(hydroxymethyl)-5-methoxyoxane-3,4-diol
SMILES COC1OC(CO)C(OC2OC(CO)C(OC)C(O)C2O)C(O)C1O

Specifications

CAS Max % 1
Quantity 100 g
Grade Multi-Compendial
Packaging Poly Bottle