missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Microcrystalline Cellulose, NF, BP, JP, Spectrum™ Chemical
C4578, 9004-34-6
Supplier: Spectrum Chemical Mfg Cor C457812KG
Description
Spectrum™ Chemical Microcrystalline Cellulose, NF, BP, JPSpecifications
| 9004-34-6 | |
| C14H26O11 | |
| PTHCMJGKKRQCBF-UHFFFAOYNA-N | |
| 2-{[4,5-dihydroxy-2-(hydroxymethyl)-6-methoxyoxan-3-yl]oxy}-6-(hydroxymethyl)-5-methoxyoxane-3,4-diol | |
| Multi-Compendial |
| 1 | |
| 12 kg | |
| COC1OC(CO)C(OC2OC(CO)C(OC)C(O)C2O)C(O)C1O | |
| 370.35 | |
| Fiber Drum |
Chemical Identifiers
| 9004-34-6 | |
| 370.35 | |
| 2-{[4,5-dihydroxy-2-(hydroxymethyl)-6-methoxyoxan-3-yl]oxy}-6-(hydroxymethyl)-5-methoxyoxane-3,4-diol |
| C14H26O11 | |
| PTHCMJGKKRQCBF-UHFFFAOYNA-N | |
| COC1OC(CO)C(OC2OC(CO)C(OC)C(O)C2O)C(O)C1O |