missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Micrococcal Nuclease (300 U/μL)

Catalog No. FEREN0181
Change view
Click to view available options
Quantity:
8,000 units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEREN0181 8,000 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEREN0181 Supplier Thermo Scientific™ Supplier No. EN0181

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Digest single-stranded and double-stranded DNA and RNA with Thermo Scientific™ Micrococcal Nuclease (S7 Nuclease), an endo-exonuclease.

Micrococcal Nuclease (S7 Nuclease) is a relatively nonspecific endo-exonuclease that digests single-stranded and double-stranded nucleic acids, but is more active on single-stranded substrates. Cleavage of DNA or RNA occurs preferentially at AT or AU-rich regions, yielding mononucleotides and oligonucleotides with terminal 3'-phosphates. The enzyme's activity is strictly dependent on Ca2+.

Applications

  • Hydrolysis of nucleic acids in crude cell-free extracts
  • Sequencing of RNA
  • Studies of chromatin structure
  • A model for protein folding and for structure-function studies

Specifications

Concentration 300 U/μL
Enzyme Nuclease
Quantity 8,000 units
Product Type Micrococcal Nuclease

For Research Use Only. Not for use in diagnostic procedures.