missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Microcentrifuge Tubes with Locking Snap Cap

Polypropylene microcentrifuge tubes are DNase/RNase and pyrogen free

 View more versions of this product

Catalog No. 14666318

Only null left

Description

Description

Polypropylene microcentrifuge tubes are DNase/RNase and pyrogen free

  • DNase/RNase and pyrogen free
  • Autoclavable and withstand temperatures to -86°C
  • Graduated tubes available in three sizes - 0.5mL, 1.5mL and 2.0mL
  • Individually wrapped
Specifications

Chemical Identifiers

Locking snap cap
Autoclavable
Polypropylene
Yes

Specifications

Locking snap cap
1.5 mL
Polypropylene
Sterile
26,000 x g
39 mm
Autoclavable
Natural
Individually wrapped
Yes
13 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions