missing translation for 'onlineSavingsMsg'
Learn More

Simport™ Scientific Micrewtube™ Microcentrifuge Tubes with Low Adhesion Surface

Micrewtube™ Microcentrifuge Tubes with Low Adhesion Surface are ideal for research procedures such as nucleic acid amplifications, protein work and others.

Supplier:  Simport™ Scientific T3414TLST

 View more versions of this product

Catalog No. 23-045-259


Only null left
Explore available promotions

Description

Description

  • Made of polypropylene
  • Autoclavable
  • Certified RNase, DNase, DNA and Pyrogen free
  • Max force: 17,000 g
  • No lubricants (such as silicone) necessary, thereby eliminating the danger of sample contamination
  • Will withstand temperatures from -196°C to +121°C
Specifications

Specifications

RNase, DNase, Pyrogen and DNA-free
Polypropylene
1.5 mL
Self-standing
Autoclavable
1000 Pk.
Non-sterile
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Accessories
Promotions

Promotions