Learn More
(±)-Miconazole nitrate salt, Thermo Scientific Chemicals
$173.43 - $173.43
Chemical Identifiers
| CAS | 22832-87-7 |
|---|---|
| Molecular Formula | C18H15Cl4N3O4 |
| Molecular Weight (g/mol) | 479.14 |
| MDL Number | MFCD00058161 |
| InChI Key | MCCACAIVAXEFAL-UHFFFAOYNA-N |
| PubChem CID | 68553 |
| IUPAC Name | 1-[2-(2,4-dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]imidazole;nitric acid |
| SMILES | O[N+]([O-])=O.ClC1=CC=C(COC(CN2C=CN=C2)C2=CC=C(Cl)C=C2Cl)C(Cl)=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC291830050
|
Thermo Scientific Chemicals
291830050 |
5 g |
Each for $173.43
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 22832-87-7 | |
| 479.14 | |
| MCCACAIVAXEFAL-UHFFFAOYNA-N | |
| 1-[2-(2,4-dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]imidazole;nitric acid |
| C18H15Cl4N3O4 | |
| MFCD00058161 | |
| 68553 | |
| O[N+]([O-])=O.ClC1=CC=C(COC(CN2C=CN=C2)C2=CC=C(Cl)C=C2Cl)C(Cl)=C1 |
Specifications
| (±)-Miconazole Nitrate Salt | |
| Authentic | |
| 5 g | |
| MFCD00058161 | |
| MCCACAIVAXEFAL-UHFFFAOYNA-N | |
| 1-[2-(2,4-dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]imidazole;nitric acid | |
| 68553 | |
| ≥97.5% (HPLC) |
| 22832-87-7 | |
| 0.5% max. (105°C, 2 h) | |
| C18H15Cl4N3O4 | |
| Solubility in water: slightly soluble. | |
| O[N+]([O-])=O.ClC1=CC=C(COC(CN2C=CN=C2)C2=CC=C(Cl)C=C2Cl)C(Cl)=C1 | |
| 479.14 | |
| 479.14 | |
| (±)-Miconazole nitrate salt |
Safety and Handling
GHS H Statement Harmful if swallowed. May cause an allergic skin reaction.
GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. If skin irritation or rash occurs: Get medical advice/attention. Wash contaminated clothing before reuse.
Warning
RUO – Research Use Only