missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MFG Tray Chemical-Resistant Multipurpose Trays and Pans
Ideal for soaking, cleaning, autoclaving, storing or carrying lab products and containing spills
Supplier: MFG Tray 3054031537
Description
Ideal for soaking, cleaning, autoclaving, storing or carrying lab products and containing spills
- Reinforced white thermoset polyester composite construction Resistant to most detergents, mild acid and alkaline solutions, corrosive salts and many reagents
- Withstand temperatures from -51° to +121°C (-60° to +250°F) continuous exposure, or 149°C (300°F) intermittent
- High load-bearing capacity
- Excellent dimensional stability Easily steam cleaned
Order Info
Available in 1 in. deep (2.5cm) trays, shallow pans from 1 1/4 to 2 3/4 in. deep (3 to 7cm), and ventilated stacking pans from 2 3/4 to 4 1/8 in. deep (7 to 10.5cm).
Specifications
| 21.88 x 16 x 1 in. | |
| 2.5 cm | |
| Fiberglass | |
| 41 cm | |
| 21.88 in. | |
| Rectangular | |
| -51°C to +121°C |
| White | |
| 55.5 cm | |
| Multi-purpose Tray | |
| Ideal for soaking, cleaning, autoclaving, storing or carrying lab products and containing spills | |
| 1 in. | |
| -60°F to +250°F | |
| 16 in. |