missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MFG Tray Chemical-Resistant Multipurpose Trays and Pans
Description
Ideal for soaking, cleaning, autoclaving, storing or carrying lab products and containing spills
- Reinforced white thermoset polyester composite construction Resistant to most detergents, mild acid and alkaline solutions, corrosive salts and many reagents
- Withstand temperatures from -51° to +121°C (-60° to +250°F) continuous exposure, or 149°C (300°F) intermittent
- High load-bearing capacity
- Excellent dimensional stability Easily steam cleaned
Order Info
Available in 1 in. deep (2.5cm) trays, shallow pans from 1 1/4 to 2 3/4 in. deep (3 to 7cm), and ventilated stacking pans from 2 3/4 to 4 1/8 in. deep (7 to 10.5cm).
Specifications
Specifications
| Dimensions (L x W x H) | 31 x 25 x 2.75 in. |
| Color | White |
| Height (Metric) | 7 cm |
| Length (Metric) | 63.5 cm |
| Material | Fiberglass |
| Type | Multi-purpose Tray and Shallow Pan |
| Width (Metric) | 63.5 cm |
| For Use With (Application) | Ideal for soaking, cleaning, autoclaving, storing or carrying lab products and containing spills |
| Length (English) | 31 in. |
| Height (English) | 2.75 in. |
| Show More |