missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ MF-™ Mixed Cellulose Ester Membranes: Triton-free

Catalog No. HATF07500
Change view
Click to view available options
Diameter (Metric):
75 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
HATF07500 75 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. HATF07500 Supplier MilliporeSigma™ Supplier No. HATF07500

Composed of mixed cellulose esters (nitrate and acetate) with minimal amounts of wetting agent. MilliporeSigma™ MF-Millipore™ Mixed Cellulose Ester Membranes: Triton-free are designed for critical applications involving sensitive biological solutions, cell contact or very small volumes.

Specifications

Autoclavable Autoclavable
Color White
Material MCE
Pore Size 0.45 μm
For Use With (Application) Clarification of fluids; sterilizing filtration of tissue culture media, additives, antibiotics; air monitoring; particulate analysis; particulate removal; biological assays (receptor binding studies, TCA precipitates, etc.).
Membrane Type Plain
Diameter (Metric) 75 mm