Learn More
MilliporeSigma™ MF-Millipore™ Mixed Cellulose Ester Membranes, Triton-free
MF-Millipore™ Mixed Cellulose Ester Membranes, Triton-free composed of mixed cellulose esters (nitrate and acetate).
Supplier: MilliporeSigma™ HATF01300
Description
Composed of mixed cellulose esters (nitrate and acetate)
Biologically inert mixtures of cellulose acetate and cellulose nitrate have made MF-Millipore™ membrane filters one of the most widely used membranes in analytical and research applications. MF-Millipore™ filters without Triton™ surfactant contain minimum amounts of wetting agent and have a lower water extractable content than standard MF-Millipore™ filters.
Features and Benefits:- Versatile filter for biological and environmental monitoring applications
- Available in a range of pore sizes, colored black or white, with or without a gridded surface
- Compatible with ethylene oxide, gamma irradiation, and autoclave sterilization methods
Specifications
| Not Autoclavable | |
| Reusable | |
| Disc | |
| 0.45 μm | |
| 60 mL/min. x cm2 | |
| Plain | |
| Hydrophilic | |
| 13 mm |
| White | |
| Membrane Filter | |
| MCE | |
| Non-sterile | |
| Clarification of fluids, Sterilizing filtration of tissue culture media, additives, antibiotics, Air monitoring, Particulate analysis, Particulate removal, Biological assays (receptor binding studies, TCA precipitates, etc.). | |
| Blotting Membranes | |
| 0.5 in. |