missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ MF-Millipore™ Mixed Cellulose Ester Membranes, Triton-free

Catalog No. HATF02500
Change view
Click to view available options
Pore Size:
0.22 μm
0.45 μm
Diameter (Metric):
13 mm
25 mm
47 mm
90 mm
142 mm
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size Diameter (Metric)
HATF02500 0.45 μm 25 mm
HATF14250 0.45 μm 142 mm
GSTF02500 0.22 μm 25 mm
HATF09025 0.45 μm 90 mm
GSTF01300 0.22 μm 13 mm
HATF04700 0.45 μm 47 mm
HATF01300 0.45 μm 13 mm
GSTF04700 0.22 μm 47 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. HATF02500 Supplier MilliporeSigma™ Supplier No. HATF02500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MF-Millipore™ Mixed Cellulose Ester Membranes, Triton-free composed of mixed cellulose esters (nitrate and acetate).

Composed of mixed cellulose esters (nitrate and acetate)

Biologically inert mixtures of cellulose acetate and cellulose nitrate have made MF-Millipore™ membrane filters one of the most widely used membranes in analytical and research applications. MF-Millipore™ filters without Triton™ surfactant contain minimum amounts of wetting agent and have a lower water extractable content than standard MF-Millipore™ filters.

Features and Benefits:
  • Versatile filter for biological and environmental monitoring applications
  • Available in a range of pore sizes, colored black or white, with or without a gridded surface
  • Compatible with ethylene oxide, gamma irradiation, and autoclave sterilization methods

Specifications

Autoclavable Not Autoclavable
Color White
Disposable Reusable
Filter Type Membrane Filter
Format Disc
Material MCE
Pore Size 0.45 μm
Sterility Non-sterile
Flow Rate 60 mL/min. x cm2
For Use With (Application) Clarification of fluids, Sterilizing filtration of tissue culture media, additives, antibiotics, Air monitoring, Particulate analysis, Particulate removal, Biological assays (receptor binding studies, TCA precipitates, etc.).
Membrane Type Plain
Type Blotting Membranes
Wettability Hydrophilic
Diameter (English) 1 in.
Diameter (Metric) 25 mm
Show More Show Less