missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Mettler Toledo™ SevenGo™ Series Portable Meter: Model SG78
Characterized by user-friendliness and excellent ergonomics. Mettler Toledo SevenGo Series SG78 Portable Meter is watertight and robust instrument with high measurement accuracy.
Manufacturer: Mettler Toledo™ 51302621
Description
- Easy and intuitive operation
- pH range: -2 through +19.99
- Large, easy-to-read display screen
- Comprehensive range of practical accessories
- Modern GLP data management
- Revolutionary Intelligent Sensor Management (ISM™)
Specifications
Meter, batteries, wrist strap, test certificate and declaration of conformity, InLab Expert-Pro-ISM-IP67 and InLab 738-ISM (both IP67 and 1.8m fixed cable) | |
0.1/0.01/0.001 | |
-1999 to 1999mV | |
±0.1mV | |
0.01/1 | |
0.01mg/L to 300g/L | |
±0.5% | |
0 to 100megohm/cm | |
0.1°C | |
LCD (Graphic with backlight) | |
IR to printer or PC via RS232 or USB | |
IP-67 | |
Determination of pH, mV, rel. mV or ion concentrations and conductivity, TDS (total dissolved solids), salinity or specific resistance | |
NTC |
-2 to 19.999 | |
±0.002 | |
0.1mV | |
0.01μS/cm to 1000mS/cm | |
±0.5% | |
0.01/1 | |
0 to 80ppt | |
5 Points, 7 pre-defined buffer group | |
±0.2°C | |
pH: BNC/RCA, conductor: LTW 7-pin | |
4 x AA | |
GLP/IP 67 | |
Electrode Kit |
Promotions
-
Get Up to 3X the Fisher Scientific Buck$ with Purchase
Buy a Mettler Toledo balance or analytical instrument and get up to three times the Fisher Scientific Buck$.: 01/31/20