missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

METTLER TOLEDO™ Portable pH/Conductivity/Dissolved Oxygen/Ion Meters

Water and dust proof, IP67 rated meter for portable or field measurements. Portable meters compatible with pH, conductivity, dissolved oxygen, and ion selective electrodes.

Supplier:  METTLER TOLEDO™ 30207958

 View more versions of this product

Catalog No. 01-913-974



Includes: Meter, electrode clip, operating instructions, quick guide, test report, declaration of conformity and batteries.

Description

Description

  • Dust proof
  • Water proof
  • IP67 Rated
  • pH, Redox/ORP, conductivity, dissolved oxygen and ISE measuring capability (model dependent)
  • Compatible with a wide range of IP67-rated electrodes for any application
  • Compatible with our ISM electrodes that capture and save calibration data which is then automatically recognized by the meter once connected
  • Data storage available; capacity dependent on model
  • Export data to a printer or Excel file
  • EasyDirect software available for data transfer (model dependent)
  • Rubber holster available for single-handed measurements
  • uGo carrying and protecting case included in selective kits (see below for complete list); available for all portable meters

Certifications

ISO 9001, ISO 14001

Specifications

Specifications

±0.2 mg/L in range 0 to 15 mg/L; ±10% in range 15 to 60 mg/L
4 x 1.5 V AA or 1.3 V NiMH acc.
200 (Memory)
Single-channel
IP 67
0°C to 60°C
Meter only
Chinese, Chinese (TW), Croatian, Czech, Danish, Dutch, English (GB), English (US), French, German, Hungarian, Indonesian, Italian, Japanese, Korean, Norwegian, Polish, Portuguese (BR), Russian, Slovak, Slovenian, Spanish, Swedish, Thai, Turkish, Vietnamese
0.1 to 600%
500 to 1100 mbar
Seven2Go S4 DO Meter
0.01
270 g
250 to 400 hr.
ISO 9001, ISO 14001
Seven2Go DO meter S4
LCD
0.1 to 600%
Robust polarographic dissolved oxygen determination outdoors.
BNC (1012 Ohm)/RCA (cinch) (sensor)
Single channel; conductivity, TDS, salinity
0.1
1
0.01 to 99.99mg/L (ppm)
222 x 70 x 35 mm
SDS
Documents

Documents

Promotions

Promotions