missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ Buffer Solutions: Standard pH
GREENER_CHOICE

Storage and refill solutions for pH electrodes

$199.28 - $236.70

Specifications

Boiling Point ∼100°C
Type Buffer Solution
Filling Solution Type pH Buffer
Includes Quality inspection certificate
Water Soluble Completely Miscible
View More Specs

Products 2
Catalog Number Mfr. No. Color pH Range Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Color pH Range Packaging Price Quantity & Availability  
01913209
GREENER_CHOICE
View Documents Promotion Details
Mettler Toledo™
51302047
Clear, Green 7 30 x 20 mL Sachets
Case of 30 Each for $236.70
Only null left
 
01915646
GREENER_CHOICE
View Documents Promotion Details
Mettler Toledo™
51302069
Clear, Red 4 30 x 20 mL Sachets
Pack of 30 Each for $199.28
Only null left
 
Description

Description

  • Completely miscible
  • Comes with Quality inspection certificate
  • NIST traceable
Specifications

Specifications

∼100°C
pH Buffer
Completely Miscible
Buffer Solution
Quality inspection certificate
Videos
SDS
Documents

Documents