missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ Buffer Solutions: Standard pH
GREENER_CHOICE

Catalog No. 01915646
Change view
Click to view available options
pH:
4
7
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH
01915646 4
01913209 7
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01915646 Supplier Mettler Toledo™ Supplier No. 51302069

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Storage and refill solutions for pH electrodes

  • Completely miscible
  • Comes with Quality inspection certificate
  • NIST traceable

Specifications

Boiling Point ∼100°C
Color Clear, Red
pH 4
Solution Type pH Buffer
Packaging 30 x 20 mL Sachets
Includes Quality inspection certificate
Solubility Information Completely Miscible in Water