missing translation for 'onlineSavingsMsg'
Learn More

Metronidazole, 99%

CAS: 443-48-1 | C6H9N3O3 | 171.156 g/mol

Supplier:  Thermo Scientific Chemicals H6025822

 View more versions of this product

Catalog No. AAH6025822


Only null left
Explore available promotions

Description

Description

Metronidazole is a limited spectrum antibiotic that actively deters the growth of protozoa, anaerobic gram-positive, and anaerobic gram-negative bacteria. The first use of this antibiotic was to suppress protozoans such as Entamoeba histolytica, Giardia lamblia and Trichomonas vaginalis. It has been used to repress the growth of gram-negative anaerobes belonging to the Bacteroides and Fusobacterium, and gram-positive anaerobes such aspeptostreptococcus and Clostridia. The advantages of this antibiotic is that it affects a high percentage of gram-negative bacteria and has greater tissue penetration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Metronidazole is a limited spectrum antibiotic that actively deters the growth of protozoa, anaerobic gram-positive, and anaerobic gram-negative bacteria. The first use of this antibiotic was to suppress protozoans such as Entamoeba histolytica, Giardia lamblia and Trichomonas vaginalis. It has been used to repress the growth of gram-negative anaerobes belonging to the Bacteroides and Fusobacterium, and gram-positive anaerobes such aspeptostreptococcus and Clostridia. The advantages of this antibiotic is that it affects a high percentage of gram-negative bacteria and has greater tissue penetration.

Solubility
Soluble in water (10 mg/ml at 20°C), ethanol, methanol, chloroform, and dimethyl sulfoxide.

Notes
Store at 4°C.
Specifications

Specifications

Metronidazole
159°C to 161°C
MFCD00009750
611683
Soluble in water (10mg/ml at 20°C),ethanol,methanol,chloroform,and dimethyl sulfoxide.
CC1=NC=C(N1CCO)[N+](=O)[O-]
171.156
CHEBI:6909
99%
443-48-1
C6H9N3O3
100 g
metronidazole, metronidazol, flagyl, 2-methyl-5-nitroimidazole-1-ethanol, anagiardil, trichazol, gineflavir, meronidal, metronidaz, novonidazol
VAOCPAMSLUNLGC-UHFFFAOYSA-N
2-(2-methyl-5-nitroimidazol-1-yl)ethanol
4173
171.16

Chemical Identifiers

443-48-1
171.156
VAOCPAMSLUNLGC-UHFFFAOYSA-N
4173
2-(2-methyl-5-nitroimidazol-1-yl)ethanol
C6H9N3O3
MFCD00009750
metronidazole, metronidazol, flagyl, 2-methyl-5-nitroimidazole-1-ethanol, anagiardil, trichazol, gineflavir, meronidal, metronidaz, novonidazol
CHEBI:6909
CC1=NC=C(N1CCO)[N+](=O)[O-]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H351
Suspected of causing cancer.

P201-P202-P281-P308+P313-P501c

H350

EINECSNumber : 207-136-1

RTECSNumber : NI5600000

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only