missing translation for 'onlineSavingsMsg'
Learn More

Metronidazole, 99%

Catalog No. p-7040478
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6025814 25 g
AAH6025822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6025814 Supplier Thermo Scientific Chemicals Supplier No. H6025814

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 443-48-1 | C6H9N3O3 | 171.156 g/mol

Metronidazole is a limited spectrum antibiotic that actively deters the growth of protozoa, anaerobic gram-positive, and anaerobic gram-negative bacteria. The first use of this antibiotic was to suppress protozoans such as Entamoeba histolytica, Giardia lamblia and Trichomonas vaginalis. It has been used to repress the growth of gram-negative anaerobes belonging to the Bacteroides and Fusobacterium, and gram-positive anaerobes such aspeptostreptococcus and Clostridia. The advantages of this antibiotic is that it affects a high percentage of gram-negative bacteria and has greater tissue penetration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Metronidazole is a limited spectrum antibiotic that actively deters the growth of protozoa, anaerobic gram-positive, and anaerobic gram-negative bacteria. The first use of this antibiotic was to suppress protozoans such as Entamoeba histolytica, Giardia lamblia and Trichomonas vaginalis. It has been used to repress the growth of gram-negative anaerobes belonging to the Bacteroides and Fusobacterium, and gram-positive anaerobes such aspeptostreptococcus and Clostridia. The advantages of this antibiotic is that it affects a high percentage of gram-negative bacteria and has greater tissue penetration.

Solubility
Soluble in water (10 mg/ml at 20°C), ethanol, methanol, chloroform, and dimethyl sulfoxide.

Notes
Store at 4°C.

Chemical Identifiers

CAS 443-48-1
Molecular Formula C6H9N3O3
Molecular Weight (g/mol) 171.156
MDL Number MFCD00009750
InChI Key VAOCPAMSLUNLGC-UHFFFAOYSA-N
Synonym metronidazole, metronidazol, flagyl, 2-methyl-5-nitroimidazole-1-ethanol, anagiardil, trichazol, gineflavir, meronidal, metronidaz, novonidazol
PubChem CID 4173
ChEBI CHEBI:6909
IUPAC Name 2-(2-methyl-5-nitroimidazol-1-yl)ethanol
SMILES CC1=NC=C(N1CCO)[N+](=O)[O-]

Specifications

Melting Point 159°C to 161°C
Quantity 25 g
Beilstein 611683
Solubility Information Soluble in water (10mg/ml at 20°C),ethanol,methanol,chloroform,and dimethyl sulfoxide.
Formula Weight 171.16
Percent Purity 99%
Chemical Name or Material Metronidazole
Hazard Category H350
Hazard Statement GHS H Statement
H351
Suspected of causing cancer.
Precautionary Statement P201-P202-P281-P308+P313-P501c
EINECSNumber 207-136-1
RTECSNumber NI5600000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only