missing translation for 'onlineSavingsMsg'
Learn More

Methyltriphenylphosphonium Chloride 98.0+%, TCI America™

Catalog No. M27025G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M27025G 5 g
M270225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M27025G Supplier TCI America Supplier No. M27025G

Chemical Identifiers

CAS 1031-15-8
Molecular Formula C19H18ClP
Molecular Weight (g/mol) 312.78
MDL Number MFCD00797851
InChI Key QRPRIOOKPZSVFN-UHFFFAOYSA-M
Synonym methyltriphenylphosphonium chloride, methyl triphenyl phosphonium chloride, phosphonium, methyltriphenyl-, chloride, methyltriphenylphosphanium chloride, methyltriphenylphosphoniumchloride, methyl triphenylphosphoniumchloride, methyl triphenylphosphonium chloride, methyl triphenyl phosphanium chloride
PubChem CID 9879809
IUPAC Name methyltriphenylphosphanium chloride
SMILES [Cl-].C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 223°C
Color White
Quantity 5 g
UN Number 3077
Formula Weight 312.78
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Methyltriphenylphosphonium Chloride