missing translation for 'onlineSavingsMsg'
Learn More

Methyltriphenylphosphonium chloride, 97%, Thermo Scientific Chemicals

$93.21 - $93.21

Chemical Identifiers

CAS 1031-15-8
Molecular Formula C19H18ClP
Molecular Weight (g/mol) 312.78
MDL Number MFCD00797851
InChI Key QRPRIOOKPZSVFN-UHFFFAOYSA-M
Synonym methyltriphenylphosphonium chloride, methyl triphenyl phosphonium chloride, phosphonium, methyltriphenyl-, chloride, methyltriphenylphosphanium chloride, methyltriphenylphosphoniumchloride, methyl triphenylphosphoniumchloride, methyl triphenylphosphonium chloride, methyl triphenyl phosphanium chloride
PubChem CID 9879809
IUPAC Name methyl(triphenyl)phosphanium;chloride
SMILES [Cl-].C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5685106
View Documents
Thermo Scientific Chemicals
H5685106
5 g
Each for $93.21
Only null left
 
Description

Description

Methyltriphenylphosphonium chloride is used as a phase transfer catalyst in synthetic chemistry. It acts as a catalyst for regio-controlled ring-opening polymerization of substituted epoxides. It is utilized for controlling the corrosion iron. It acts as a reactant for intramolecular nitrone cycloaddition. In addition to this, it is used for neoplasm inhibition through antimitochondrial effect.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyltriphenylphosphonium chloride is used as a phase transfer catalyst in synthetic chemistry. It acts as a catalyst for regio-controlled ring-opening polymerization of substituted epoxides. It is utilized for controlling the corrosion iron. It acts as a reactant for intramolecular nitrone cycloaddition. In addition to this, it is used for neoplasm inhibition through antimitochondrial effect.

Solubility
Soluble in methanol.

Notes
Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

1031-15-8
312.78
QRPRIOOKPZSVFN-UHFFFAOYSA-M
9879809
[Cl-].C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
C19H18ClP
MFCD00797851
methyltriphenylphosphonium chloride, methyl triphenyl phosphonium chloride, phosphonium, methyltriphenyl-, chloride, methyltriphenylphosphanium chloride, methyltriphenylphosphoniumchloride, methyl triphenylphosphoniumchloride, methyl triphenylphosphonium chloride, methyl triphenyl phosphanium chloride
methyl(triphenyl)phosphanium;chloride

Specifications

1031-15-8
C19H18ClP
MFCD00797851
UN3077
methyltriphenylphosphonium chloride, methyl triphenyl phosphonium chloride, phosphonium, methyltriphenyl-, chloride, methyltriphenylphosphanium chloride, methyltriphenylphosphoniumchloride, methyl triphenylphosphoniumchloride, methyl triphenylphosphonium chloride, methyl triphenyl phosphanium chloride
QRPRIOOKPZSVFN-UHFFFAOYSA-M
methyl(triphenyl)phosphanium;chloride
9879809
97%
221°C (decomposition)
CH3P(C6H5)3Cl
5 g
Hygroscopic
Soluble in methanol.
[Cl-].C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
312.78
312.78
Methyltriphenylphosphonium chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318-H302-H312-H315
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.

P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c

H302+H312-H315-H318

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

EINECSNumber : 418-400-2

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only